Isotemp™ Hotplate, Ambient to 540°C, Ceramic
Isotemp™ Hotplate, Ambient to 540°C, Ceramic
Actual product may vary
Isotemp™ Hotplate, Ambient to 540°C, Ceramic
Shape: Square, Size: 4.25 x 4.25 in., Plug Type: EU and UK
Have Questions?
Catalog NumberTop Plate Material
HP88854205Ceramic
Catalog number HP88854205
Price (MYR)
2,795.00
EA
Add to cart
Top Plate Material:
Ceramic
Price (MYR)
2,795.00
EA
Add to cart
Experience exceptional safety and temperature performance for your routine protocols. Available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology:
  • Strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design:
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
  • EU and UK plug
Specifications
DescriptionHotplate
Top Plate MaterialCeramic
Size4.25 x 4.25 in.
Temperature (Metric)Ambient to 540°C
Height (English)3.6 in.
Length (English)10 in.
Width (English)5 in.
Height (Metric)9.1 cm
Length (Metric)25.4 cm
Width (Metric)12.7 cm
Electrical Requirements230 V, 50/60 Hz
Frequency50/60 Hz
Plug TypeUK/EU/CN
ShapeSquare
Voltage230 V
TypeHot Plate
Unit SizeEA