Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.
Have Questions?
Catalog Number
Grade
HGS1106
9 to 15
Catalog number HGS1106
Price (MYR)
-
Grade:
9 to 15
Learn the critical skill of micropipetting
Hands-on pipetting practice
Understand the importance of micropipetting in science
Includes instructions, fixed volume micropipette, tips, dyes, and pipetting sheet
Specifications
Grade9 to 15
Age14 to 20
For Use With (Application)At Home Education
IncludesInstructions, Fixed Volume Micropipette, Tips, Dyes, and Pipetting Sheet