Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.
Have Questions?
Catalog Number
Grade
HGS1104
6 to 15
Catalog number HGS1104
Price (MYR)
-
Grade:
6 to 15
Learn about living organisms and DNA
Hands-on experiment using real DNA
Spool DNA and visualize genetic material
Includes chromosomal DNA, spooling sticks, transfer pipettes, DNA stain
Alcohol included upon request
Specifications
DescriptionSpooling DNA on a Stick
Grade6 to 15
Age12 to 20
For Use With (Application)At Home Education
IncludesChromosomal DNA, Spooling Sticks, Transfer Pipettes, DNA Stain