MLA™ Silver Brand Precision Pipetters
MLA™ Silver Brand Precision Pipetters
Actual product may vary
MLA™ Silver Brand Precision Pipetters
Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
HGS110215 μL
HGS11015 μL
Catalog number HGS1102
Price (MYR)
-
Volume (Metric):
15 μL
  • 'To deliver' capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Capacity engraved on bonnet
Specifications
Accuracy±5%
AutoclavableNot Autoclavable
ColorSilver
DisposableReusable
For Use WithMicropipette Tip
MaterialAluminum Alloy, Stainless Steel
ModelMLA Precision
Number of Channels1
SterilityNon-sterile
TypePrecision Pipetter
Volume (Metric)15 μL
IncludesNozzle Insert, Calibration Key
Precision±1.73%
Quantity1 Ea.
WarrantyLimited Lifetime
Length (English)9 in.
Width (English)2 in.
Height (English)1.5 in.
Unit SizeCase