Scents and Sense-ability Kit
Scents and Sense-ability Kit
Actual product may vary
Scents and Sense-ability Kit
Allows students to understand that olfactory receptors respond to smells and transmit them as signals to the brain. Furthermore, students will also be able to understand the principles of thin layer chromatography and how they apply to the separation of olfactory compounds.
Have Questions?
Catalog number HGS1100
Price (MYR)
-
  • Storage: Room Temperature
  • For 10 Lab Groups
Specifications
Product TypeKit
Grade9 to Undergraduate
Age14 to 20
IncludesInstructions, Limonene, Anethole, Eugenol, S-Citronellal, Unknown Sample, Anhydrous Sodium Carbonate, Potassium Permanganate, TLC Paper, Transfer Pipets, Microcentrifuge Tubes
RequiresPencils, Beakers (100 mL, 400 mL), Gloves, Rulers, Saran Wrap, 100% Ethanol (200 or 190 Proof), and Distilled Water
Class Size10 Lab Groups
Unit SizeCase
Contents & Storage
Room Temperature