Barnstead™ Classic Still Storage Accessories
Barnstead™ Classic Still Storage Accessories
Actual product may vary
Barnstead™ Classic Still Storage Accessories
Complete your distillation system with Thermo Scientific™ Barnstead™ Classic Still Storage Accessories.
Have Questions?
Catalog NumberType
H3111Air Filter and Water Seal
Catalog number H3111
Price (MYR)
7,918.00
EA
Add to cart
Type:
Air Filter and Water Seal
Price (MYR)
7,918.00
EA
Add to cart
Thermo Scientific™ Barnstead™ Classic Still Storage Accessories complement your current system to create a more complete system. Accessories include mounting options, UV light assembly, Thermo Scientific™ Ventgard™ air filter and water seal. Choose either a floor stand or wall bracket as needed. Maintain sterility in the reservoir with the UV light assembly, which includes housing, on/off switch, 6' cord, lamp and ballast. Protect stored water against airborne particulates, organics and CO2 with Ventgard™ air filter and water seal. Prevent air from entering the reservoir via the condenser atmospheric vent.
Specifications
For Use With (Equipment)Classic Reservoir
TypeAir Filter and Water Seal
Unit SizeEA