Raised Shelf for Unstirred Water Baths
Raised Shelf for Unstirred Water Baths
Actual product may vary
Raised Shelf for Unstirred Water Baths
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
Have Questions?
Catalog NumberType
GRARS38Shelf for 34 L Tanks
Catalog number GRARS38
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Shelf for 34 L Tanks
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
  • Choose from a variety of sizes to fit your tank.
  • For use with either Stainless Steel and Plastic tank models
Specifications
For Use With (Equipment)Grant Heated Circulating Baths
TypeShelf for 34 L Tanks
Height (Metric)45 or 135 mm
Unit SizeEA