Owl™ JGC-4 Gel Casting System Replacement Parts
Owl™ JGC-4 Gel Casting System Replacement Parts
Actual product may vary
Owl™ JGC-4 Gel Casting System Replacement Parts
Maintain the quality of your current equipment with Thermo Scientific™ Owl™ JGC-4 Gel Casting System Replacement Parts, for use with the Owl™ JGC-4 Gel Casting System.
Have Questions?
Catalog NumberTypeQuantity
GP4-25Gel Pouch25 Pk.
Catalog number GP4-25
Price (MYR)
184.00
EA
Add to cart
Type:
Gel Pouch
Quantity:
25 Pk.
Price (MYR)
184.00
EA
Add to cart
Maintain the quality of your current lab equipment with Thermo Scientific™ Owl™ JGC-4 Gel Casting System Replacement Parts, for use with the Owl™ JGC-4 Gel Casting System.
  • A pouch is used to collect the excess polyacrylimide and seal the bottom of the gel.
Specifications
For Use With (Equipment)Owl JGC-4 Gel Casting System
Product LineOwl
TypeGel Pouch
Quantity25 Pk.
Unit SizeEA