Cover for Precision™ Water Baths
Cover for Precision™ Water Baths
Actual product may vary
Cover for Precision™ Water Baths
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
FTNGPSSL20Stainless Steel Gable CoverPrecision 20L and 28L General Purpose Water Baths
FISGPSSL20Stainless Steel Gable CoverPrecision 20L and 28L General Purpose Water Baths
Catalog number FTNGPSSL20
Price (MYR)
2,555.00
EA
Add to cart
Description:
Stainless Steel Gable Cover
For Use With (Equipment):
Precision 20L and 28L General Purpose Water Baths
Price (MYR)
2,555.00
EA
Add to cart
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Gable Covers
  • Enhanced hinged design supports open lid configuration and easy lid removal
  • Accommodate taller labware with extended height
  • Polycarbonate and stainless steel models available
  • Warranty varies by country, please contact us for details
Specifications
DescriptionStainless Steel Gable Cover
Capacity (Metric)20, 28 L
For Use With (Equipment)Precision 20L and 28L General Purpose Water Baths
TypeGable Cover