Isotemp™ General Purpose Deluxe Water Baths
Isotemp™ General Purpose Deluxe Water Baths
Actual product may vary
Isotemp™ General Purpose Deluxe Water Baths
Choose general purpose water baths, designed to provide consistent temperature for reliable results with easy-to-use operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelCapacity (Metric)Dimensions (D x W x H) InteriorDimensions (L x W x H)
FTNFSGPD2SIsotemp GPD 2S2 L6 x 11.8 x 2.69 in. (153 x 300 x 65 mm)9.7 x 14 x 9.1 in. (246 x 355 x 232 mm)
FSGPD02Isotemp GPD 022 L5.4 x 6.1 x 5.99 in. (138 x 155 x 150 mm)9.1 x 7.8 x 9.2 in. (230 x 199 x 233 mm)
FSGPD05Isotemp GPD 055 L6.1 x 11.8 x 5.99 in. (154 x 300 x 150 mm)9.7 x 14 x 9.1 in. (246 x 355 x 232 mm)
FSGPD10Isotemp GPD 1010 L11.9 x 13 x 5.99 in. (301 x 330 x 150 mm)15.5 x 15.1 x 9.2 in. (393 x 383 x 233 mm)
FSGPD20Isotemp GPD 2020 L11.7 x 19.7 x 5.99 in. (297 x 500 x 150 mm)15.4 x 21.8 x 9.2 in. (392 x 555 x 233 mm)
FKS15305887Isotemp GPD 2828 L11.7 x 19.7 x 7.99 in. (297 x 500 x 200 mm)15.4 x 21.8 x 11.1 in. (392 x 555 x 282 mm)
FSGPD28Isotemp GPD 2828 L11.7 x 19.7 x 7.99 in. (297 x 500 x 200 mm)15.4 x 21.8 x 11.1 in. (392 x 555 x 282 mm)
Catalog number FTNFSGPD2S
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Model:
Isotemp GPD 2S
Capacity (Metric):
2 L
Dimensions (D x W x H) Interior:
6 x 11.8 x 2.69 in. (153 x 300 x 65 mm)
Dimensions (L x W x H):
9.7 x 14 x 9.1 in. (246 x 355 x 232 mm)
Ideal for industrial, clinical pharmaceutical or biomedical applications, Fisherbrand™ Isotemp™ general purpose deluxe water baths provide powerful technology in a compact design. Available in a variety of sizes to meet a wide range of application requirements, the Isotemp water baths are designed to provide consistent temperature for reliable results with easy-to-use operation.
  • Smaller footprint, compared to previous models
  • Advanced temperature controller is designed for extended functionality 
  • New audible alarms
  • Conveniently save commonly used settings with temperature presets
  • Simple to operate and monitor with icon-based graphical display
  • Enhanced hinge design supports open lid configuration and easy lid removal
  • Help prevent bath damage and overheating with low-fluid protection
  • Compatable with Thermal beads (sold separately)
Specifications
Chamber MaterialStainless Steel
Controller TypeDigital
Description2L Shallow Deluxe General Purpose Water Bath
Capacity (Metric)2 L
Certifications/ComplianceUL, CE, FDA Class I
Temperature RangeAmbient to 100°C
DisplayMonochrome LCD
Heating Capacity300W
IncludesPolycarbonate Gable Cover, Diffuser Tray
MaterialEpoxy Powder-coated Steel
ModelIsotemp GPD 2S
Temperature Presets3
Temperature Stability±0.2°C
Temperature Uniformity±0.2°C
Dimensions (D x W x H) Interior6 x 11.8 x 2.69 in. (153 x 300 x 65 mm)
Dimensions (L x W x H)9.7 x 14 x 9.1 in. (246 x 355 x 232 mm)
Amperage2.2-2.7 / 1.0-1.3 (120V/230V)
Unit SizeEach