accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
Actual product may vary
accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
accumet™ Solid State Combination ISEs, Mercury-Free: Fluoride
For fluoride determinations.
Have Questions?
Catalog NumberConnector Types
FSP13620629BNC
Catalog number FSP13620629
Price (MYR)
-
Connector Types:
BNC
For fluoride determinations.
Features
  • Low drift, fast response, high selectivity ratio, good thermal stability and wide operating range
  • Continuous electrical shielding and insulation of internal elements, cable and connector
  • Extremely stable, reproducible readings with a minimum of electrical noise
  • Rapid response time that lowers cost per test
  • Individually tested, serialized to meet GLP requirements and backed by the Fisher Scientific Technical Applications staff
Combination models
  • Feature a homogenous sensing membrane for wide application, rapid response, minimum sample retention, improved precision, and lower detection limits
  • In glass or epoxy body incorporating sensing and reference cells in a single probe
Fluoride Combination ISE
  • Determines F- concentration down to 1 × 10-6M (0.02ppm)
  • Applications include analyses of water, wastewater, stack gases, explosives, etching baths, research with biological fluids and tissues, beverages, detergents
  • Method has ASTM and EPA approval
  • Glass body
Specifications
ApplicationsUsed to analyse water, wastewater, stack gases, explosives, etching baths, research with biological fluids and tissues, beverages and detergents
Connector TypesBNC
Diameter (English) Body0.5 in.
Diameter (Metric) Body13mm
Filling Solution TypeSP138-500
Length (English)4.25 in.
Length (Metric)108mm
MaterialGlass
Operating Range (Metric)0.02 to saturationppm
Temperature (English) Operating32° to 176°F
pH Range5 to 8
InterferencesOH-
ModelFluoride Combination
ElectrodeFluoride
Temperature (Metric) Range0 to 80°C
Temperature (Metric) Operating0°C to 80°C
TypeCombination ISE
Unit SizeEach