accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
Actual product may vary
accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeProbe TypeMaterial
FSP13-636-AB315ApH/mV Meter with ProbeGel-filled pH/ATC ElectrodeEpoxy Body
FSP13-636-AB315pH/mV Meter
FSP13-636-AB315BpH/mV Meter with ProbeRefillable pH ElectrodeGlass Body
Catalog number FSP13-636-AB315A
Price (MYR)
4,180.00
Special offer
Ends: 31-Dec-2025
5,726.00
Save 1,546.00 (27%)
EA
Add to cart
Type:
pH/mV Meter with Probe
Probe Type:
Gel-filled pH/ATC Electrode
Material:
Epoxy Body
Price (MYR)
4,180.00
Special offer
Ends: 31-Dec-2025
5,726.00
Save 1,546.00 (27%)
EA
Add to cart
  • Large, backlit display simultaneous shows pH, mV and temperature readings
  • One to five point pH calibration with automatic buffer recognition of four pH buffer sets
  • One point relative mV calibration (RmV) for ORP measurements
  • One point ATC temperature probe calibration
  • Calibration due alarm, selectable 1 to 60 hr., or off
  • Automatic or manual temperature compensation
  • Ready icon indicates measurement stability
  • Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
  • Non-volatile meter memory preserves settings and data in event of power loss
  • Meter-attached stand facilitates easy electrode movement into and out of samples
Specifications
Accuracy (Temperature)±0.3°C (±0.5°F)
Accuracy (mV)±0.2 or ±0.05%, Whichever is Greater
Accuracy (pH)±0.01
Data Management DetailsPrinted or CSV Format
DescriptionpH/mV Meter with pH/ATC Epoxy-body Double Junction Gel-filled Electrode
OutputsPort for Computer or Printer
IncludespH/ATC Electrode, Electrode Stand, Power Adapter
Measurement ModespH with mV and Temperature, Relative mV with mV and Temperature
CalibrationpH, Relative mV, Temperature
Range (pH)-2 to 18
Resolution (pH)0.01, 0.1
Calibration Points (pH)USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13
Range (mV)±2000 mV
Resolution (mV)1 mV, 0.1 RmV
Calibration Points1 to 5 (pH), 1 (Relative mV), 1 (Temperature)
Temperature Range (Metric)-5°C to +105°C
Temperature Range (English)23°F to 221°F
Resolution (Temperature)0.1°C (0.1°F)
Display Type5 in. LCD
Backlight OptionYes
MemoryNon-volatile
Data Points500
Log Function TypeAutomatic, Manual
InputsBNC, ATC
Certifications/ComplianceCE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate
IP RatingIP54
AC AdapterUniversal Power Adapter
Warranty3 Years
MaterialEpoxy Body
ModelAB315
Probe TypeGel-filled pH/ATC Electrode
Range (Relative mV)±2000 mV
StabilityIndicator Icons
Temperature CompensationAutomatic, Manual
Channels1
TypepH/mV Meter with Probe
Voltage100/240 V