Posaconazole
Thermo Scientific Chemicals
Posaconazole
CAS: 171228-49-2 | C37H42F2N8O4, C37H42F2N8O4 | 700.79 g/mol
Have Questions?
Catalog NumberQuantity
FSA4676600101 g
Catalog number FSA467660010
Price (MYR)
909.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
909.00
EA
Add to cart
Specifications
Chemical Name or MaterialPosaconazole
CAS171228-49-2
Quantity1 g
Melting Point170.0°C to 179.0°C
Health Hazard 1GHS Signal Word: Warning