Phosphoric acid, for HPLC
Phosphoric acid, for HPLC
Thermo Scientific Chemicals
Phosphoric acid, for HPLC
CAS: 7664-38-2 | H3O4P | 97.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA463865000
Price (MYR)
512.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
512.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhosphoric acid
CAS7664-38-2
Melting Point42.0°C
Health Hazard 1GHS Signal Word: Danger
Density1.7000 g/mL