2-(tert-Butyl)-4,6-dimethylphenol, 97%
2-(tert-Butyl)-4,6-dimethylphenol, 97%
2-(tert-Butyl)-4,6-dimethylphenol, 97%
2-(tert-Butyl)-4,6-dimethylphenol, 97%
Thermo Scientific Chemicals
2-(tert-Butyl)-4,6-dimethylphenol, 97%
CAS: 1879-09-0 | C12H18O | 178.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA462570100
Price (MYR)
290.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
290.00
EA
Add to cart
Specifications
Chemical Name or Material2-(tert-Butyl)-4,6-dimethylphenol
Melting Point22.0°C to 23.0°C
CAS1879-09-0
Health Hazard 2GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Toxic to aquatic
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Remove/Take off immediately all