Chlorothiazide, 98%
Chlorothiazide, 98%
Chlorothiazide, 98%
Thermo Scientific Chemicals
Chlorothiazide, 98%
CAS: 58-94-6 | C7H6ClN3O4S2 | 295.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA461740050
Price (MYR)
318.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
318.00
EA
Add to cart
Specifications
Chemical Name or MaterialChlorothiazide
CAS58-94-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protecti