Irinotecan hydrochloride trihydrate
Irinotecan hydrochloride trihydrate
Irinotecan hydrochloride trihydrate
Irinotecan hydrochloride trihydrate
Thermo Scientific Chemicals
Irinotecan hydrochloride trihydrate
CAS: 136572-09-3 | HCl·3H2O | 677.18 g/mol
Have Questions?
Catalog NumberQuantity
FSA461542500250 mg
Catalog number FSA461542500
Price (MYR)
747.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
747.00
EA
Add to cart
Specifications
Chemical Name or MaterialIrinotecan hydrochloride trihydrate
CAS136572-09-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.