Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
FSA4612600101 g
Catalog number FSA461260010
Price (MYR)
486.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
486.00
EA
Add to cart
Specifications
Chemical Name or MaterialTerbinafine hydrochloride
CAS78628-80-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement Avoid release to the environment. Collect spillage. Dispose of contents/container to an approved waste disposal plant.