Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA461170010
Price (MYR)
425.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
425.00
EA
Add to cart
Specifications
Chemical Name or MaterialOseltamivir phosphate
CAS204255-11-8
Health Hazard 1Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Health Hazard 2Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation
ColorWhite to Off-White