Cortisone, 98%
Cortisone, 98%
Cortisone, 98%
Cortisone, 98%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA460520010
Price (MYR)
244.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
244.00
EA
Add to cart
Specifications
Chemical Name or MaterialCortisone
CAS53-06-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of damaging the unborn child.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.