Gatifloxacin, 98%, Thermo Scientific Chemicals
Gatifloxacin, 98%, Thermo Scientific Chemicals
Gatifloxacin, 98%, Thermo Scientific Chemicals
Gatifloxacin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gatifloxacin, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
FSA4602300101 g
Catalog number FSA460230010
Price (MYR)
237.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
237.00
EA
Add to cart
Specifications
Chemical Name or MaterialGatifloxacin
CAS112811-59-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position