Mycophenolic acid, 98%
Mycophenolic acid, 98%
Mycophenolic acid, 98%
Mycophenolic acid, 98%
Thermo Scientific Chemicals
Mycophenolic acid, 98%
CAS: 24280-93-1 | C17H20O6 | 320.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA459380050
Price (MYR)
739.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
739.00
EA
Add to cart
Specifications
Chemical Name or MaterialMycophenolic acid
CAS24280-93-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May damage the unborn child.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long last
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Use personal protective equipment as required.
IF exposed or concerned: Get m