Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals
Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals
Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals
Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA459251000
Price (MYR)
638.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
638.00
EA
Add to cart
Specifications
Chemical Name or MaterialTheophylline-7-acetic acid
CAS652-37-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee