Betamethasone, 97%
Betamethasone, 97%
Betamethasone, 97%
Thermo Scientific Chemicals
Betamethasone, 97%
CAS: 378-44-9 | C22H29FO5 | 392.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number FSA459070250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialβmethasone
CAS378-44-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.