Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Thermo Scientific Chemicals
Iodomethane, 99%, stabilized
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA459015000
Price (MYR)
867.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
867.00
EA
Add to cart
Specifications
Chemical Name or MaterialIodomethane
CAS74-88-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
CAS7440-50-8