Lovastatin, 98%
Lovastatin, 98%
Lovastatin, 98%
Thermo Scientific Chemicals
Lovastatin, 98%
Lovastatin, CAS # 75330-75-5, is a cholesterol lowering compound and inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. | CAS: 75330-75-5 | C24H36O5 | 404.55 g/mol
Have Questions?
Catalog NumberQuantity
FSA45883025025 g
Catalog number FSA458830250
Price (MYR)
619.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
619.00
EA
Add to cart
Specifications
Chemical Name or MaterialLovastatin
CAS75330-75-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
Suspected of damaging the unborn child.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
View more
General Description
Applications
• It blocks product ion of mevalonate, a critical compound in the production of cholesterol and isoprenoids
• It can increase cellular resistance to anticancer agents such as doxorubicin • It can induce apoptosis in myeloma cells
• It can cause cell cycle arrest in G1 and G2/M phase
• It also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF
RUO – Research Use Only