Ammonium hydroxide, extra pure, 10 wt% in water
Ammonium hydroxide, extra pure, 10 wt% in water
Ammonium hydroxide, extra pure, 10 wt% in water
Ammonium hydroxide, extra pure, 10 wt% in water
Thermo Scientific Chemicals
Ammonium hydroxide, extra pure, 10 wt% in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA458680025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hydroxide
CAS1336-21-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Avoid release to the environment.