Trimethylsilanol
Trimethylsilanol
Trimethylsilanol
Thermo Scientific Chemicals
Trimethylsilanol
CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA458570250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTrimethylsilanol
CAS1066-40-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.