Fusidic acid, 98%, Thermo Scientific Chemicals
Fusidic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fusidic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA458460050
Price (MYR)
2,274.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,274.00
EA
Add to cart
Specifications
Chemical Name or MaterialFusidic acid
CAS6990-06-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.