Fusidic acid, 98%, Thermo Scientific Chemicals
Fusidic acid, 98%, Thermo Scientific Chemicals
Fusidic acid, 98%, Thermo Scientific Chemicals
Fusidic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fusidic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA458460010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFusidic acid
CAS6990-06-3
Health Hazard 1Warning
Health Hazard 2GHS H Statement Harmful if swallowed.
Health Hazard 3GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.