2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™
2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™
2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™
2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™
Thermo Scientific Chemicals
2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA458070010
Price (MYR)
478.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
478.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methylpyrrolidine hydrochloride
CAS54677-53-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee