Amiodarone hydrochloride, Thermo Scientific™
Amiodarone hydrochloride, Thermo Scientific™
Amiodarone hydrochloride, Thermo Scientific™
Amiodarone hydrochloride, Thermo Scientific™
Amiodarone hydrochloride, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA457980250
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmiodarone hydrochloride
CAS19774-82-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position