3-Acetylpyridine adenine dinucleotide, 90%
3-Acetylpyridine adenine dinucleotide, 90%
3-Acetylpyridine adenine dinucleotide, 90%
3-Acetylpyridine adenine dinucleotide, 90%
Thermo Scientific Chemicals
3-Acetylpyridine adenine dinucleotide, 90%
CAS: 86-08-8 | C22H28N6O14P2 | 662.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number FSA457682500
Price (MYR)
3,817.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
3,817.00
EA
Add to cart
Specifications
Chemical Name or Material3-Acetylpyridine adenine dinucleotide
CAS86-08-8
EINECS Number201-649-4
Infrared SpectrumAuthentic
Assay Percent Range89% min. (HPLC)