Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Thermo Scientific Chemicals
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA457070010
Price (MYR)
1,069.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,069.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetrahydrofuran
Melting Point-108.4°C
CAS109-99-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement Highly flammable liquid and vapor. Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May form explosive peroxides.