Gemcitabine, 98%
Gemcitabine, 98%
Gemcitabine, 98%
Thermo Scientific Chemicals
Gemcitabine, 98%
CAS: 95058-81-4 | C9H11F2N3O4 | 263.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA456890050
Price (MYR)
1,530.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,530.00
EA
Add to cart
Specifications
Chemical Name or MaterialGemcitabine
CAS95058-81-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure if inhaled.<br
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water