Acetic acid, 25%, solution in water
Acetic acid, 25%, solution in water
Acetic acid, 25%, solution in water
Acetic acid, 25%, solution in water
Thermo Scientific Chemicals
Acetic acid, 25%, solution in water
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
Catalog number FSA456850050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic acid
CAS64-19-7
Melting Point-17.0°C
Boiling Point103.0°C
Health Hazard 1Danger