Cypermethrin, 96%
Cypermethrin, 96%
Cypermethrin, 96%
Thermo Scientific Chemicals
Cypermethrin, 96%
CAS: 52315-07-8 | C22H19Cl2NO3 | 416.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA456311000
Price (MYR)
3,320.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
3,320.00
EA
Add to cart
Specifications
Chemical Name or MaterialCypermethrin
CAS52315-07-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and