Rosuvastatin, calcium salt, 98%
Rosuvastatin, calcium salt, 98%
Rosuvastatin, calcium salt, 98%
Rosuvastatin, calcium salt, 98%
Thermo Scientific Chemicals
Rosuvastatin, calcium salt, 98%
CAS: 147098-20-2 | C22H28FN3O61/2Ca | 500.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Catalog number FSA456232500
Price (MYR)
923.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
923.00
EA
Add to cart
Specifications
Chemical Name or MaterialRosuvastatin, calcium salt
CAS147098-20-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of damaging the unborn child.
May cause harm to breast-fed children.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid contact during