Lutein, 90%
Lutein, 90%
Lutein, 90%
Thermo Scientific Chemicals
Lutein, 90%
CAS: 127-40-2 | C40H56O2 | 568.87 g/mol
Have Questions?
Catalog NumberQuantity
FSA4561600101 g
Catalog number FSA456160010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLutein
CAS127-40-2
EINECS Number204-840-
Infrared SpectrumAuthentic
Assay Percent Range88% min. (HPLC)