Oxamyl, 95%, Thermo Scientific™
Product Image
Thermo Scientific Chemicals
Oxamyl, 95%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA456140010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialOxamyl
CAS23135-22-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Fatal if swallowed.
Fatal if inhaled.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.