1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
Thermo Scientific Chemicals
1,10-Phenanthroline-5,6-dione, 98%
CAS: 27318-90-7 | C12H6N2O2 | 210.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA456080010
Price (MYR)
768.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
768.00
EA
Add to cart
Specifications
Chemical Name or Material1,10-Phenanthroline-5,6-dione
CAS27318-90-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.