Pyrimidine-2-carboxylic acid, 95%
Pyrimidine-2-carboxylic acid, 95%
Pyrimidine-2-carboxylic acid, 95%
Pyrimidine-2-carboxylic acid, 95%
Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%
CAS: 31519-62-7 | C5H4N2O2 | 124.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA455830050
Price (MYR)
1,259.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,259.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyrimidine-2-carboxylic acid
CAS31519-62-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.