Ofloxacin, 98%, Thermo Scientific Chemicals
Ofloxacin, 98%, Thermo Scientific Chemicals
Ofloxacin, 98%, Thermo Scientific Chemicals
Ofloxacin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ofloxacin, 98%, Thermo Scientific Chemicals
Ofloxacin, CAS # 82419-36-1, is a fluoroquinolone antibiotic with broad antibacterial activity against gram-positive and -negative bacteria, as well as against mycobacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number FSA455670250
Price (MYR)
1,683.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,683.00
EA
Add to cart
Specifications
Chemical Name or MaterialOfloxacin
TypeOfloxacin
CAS82419-36-1
Melting Point280.0°C to 275.0°C
Infrared SpectrumAuthentic
View more
General Description
  • Ofloxacin is a synthetic fluoroquinolone antibiotic that is an analog of nalidixic acid
  • Ofloxacin can directly interact with pneumococcal autolysin (LytA) at its choline binding domain, thus inhibiting the ability of LytA to degrade the pneumococcal cell wall
  • Ofloxacin is racemic and its (S)-enantiomer is levofloxacin
Applications
  • Ofloxacin may be used in pharmacokinetic laboratory studies and high-throughput drug screens
  • The DNA and protein-binding capabilities of ofloxacin have been studied in the laboratory
RUO – Research Use Only
General References:
  1. Fernández-Tornero, C.; García, E.; de Pascual-Teresa, B.; López, R.; Giménez-Gallego, G.; Romero, A. Ofloxacin-like antibiotics inhibit pneumococcal cell wall-degrading virulence factors. J Biol Chem. 2005, 280(20), 19948-57.
  2. Maia, A.S.; Tiritan, M.E.; Castro, P.M.L. Enantioselective degradation of ofloxacin and levofloxacin by the bacterial strains Labrys portucalensis F11 and Rhodococcus sp. FP1. Ecotoxicol Environ Saf. 2018, 155, 144-151.