Arbutin, 98%
Arbutin, 98%
Arbutin, 98%
Thermo Scientific Chemicals
Arbutin, 98%
CAS: 497-76-7 | C12H16O7 | 272.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA455640250
Price (MYR)
809.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
809.00
EA
Add to cart
Specifications
Chemical Name or MaterialArbutin
CAS497-76-7
Melting Point195.0°C to 201.0°C
EINECS Number207-850-3
Loss on Drying2% max.