Rifampin, 95%
Rifampin, 95%
Rifampin, 95%
Thermo Scientific Chemicals
Rifampin, 95%
Rifampin (rifampicin), CAS # 13292-46-1, is a synthetic rifamycin antibiotic that works by inhibiting RNA and DNA synthesis and exhibits activity against the tuberculosis microbe.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA455620050
Price (MYR)
1,205.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,205.00
EA
Add to cart
Specifications
Chemical Name or MaterialRifampin
TypeRifampicin
CAS13292-46-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
View more
General Description
• Rifampin is a rifamycin class antibiotic synthesized to resemble the compound isolated from Amycolatopsis rifamycinica
Applications
• Rifampin can be used as a bacteriostatic that prevents RNA and DNA synthesis, particularly in mycobacteria
• It may also act as an antineoplastic, a neuroprotective, and inhibitor of angiogenesis
RUO – Research Use Only
General References:
  1. Tarakji, K.G.; Mittal, S.; Kennergren, C.; Corey, R.; Poole, J.E.; Schloss, E.; Gallastegui, J.; Pickett, R.A.; Evonich, R.; Philippon, F.; McComb, J.M., Roark, S.F.; Sorrentino, D.; Sholevar, D.; Cronin, E.; Berman, B., Riggio, D.; Biffi, M.; Khan, H.; Silver, M.T.; Collier, J.; Eldadah, Z.; Wright, D.J.; Lande, J.D.; Lexcen, D.R.; Cheng, A.; Wilkoff, B.L. WRAP-IT Investigators. Antibacterial Envelope to Prevent Cardiac Implantable Device Infection. N Engl J Med. 2019, 380(20), 1895-1905.
  2. Swindells, S.; Ramchandani, R.; Gupta, A.; Benson, C.A.; Leon-Cruz, J.; Mwelase, N.; Jean Juste, M.A.; Lama, J.R.; Valencia, J.; Omoz-Oarhe, A.; Supparatpinyo, K.; Masheto, G.; Mohapi L, da Silva Escada, R.O.; Mawlana, S.; Banda, P.; Severe, P.; Hakim, J.; Kanyama, C.; Langat, D.; Moran, L.; Andersen, J.; Fletcher, C.V.; Nuermberger, E.; Chaisson, R.E., BRIEF TB/A5279 Study Team. One Month of Rifapentine plus Isoniazid to Prevent HIV-Related Tuberculosis. N Engl J Med. 2019, 380(11), 1001-1011.