Nystatin, 85+%
Nystatin, 85+%
Nystatin, 85+%
Nystatin, 85+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA455500250
Price (MYR)
1,288.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,288.00
EA
Add to cart
Specifications
Chemical Name or MaterialNystatin
TypeNystatin
CAS1400-61-9
Melting Point>160.0°C
Infrared SpectrumAuthentic