Tobramycin, 97%
Tobramycin, 97%
Tobramycin, 97%
Tobramycin, 97%
Tobramycin, CAS# 32986-56-4, is an aminoglycoside antibiotic isolated from Streptomyces tenebrarius with a wide range of bacteriostatic activity, mainly against gram-negative bacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA455430010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTobramycin
TypeTobramycin
CAS32986-56-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
View more
General Information
• Tobramycin is an aminoglycoside compound derived from kanamycin B and displays antibiotic and bacteriostatic activity against gram-negative bacteria
• After entering the cells, it binds irreversibly to a specific aminoglycoside receptor on the bacterial 30S ribosomal subunit and interferes with the initiation complex between messenger RNA and the 30S subunit, consequently inhibiting the protein synthesis process leading to bacterial cell death
• Tobramycin also can interfere with protein elongation since it can induce misreading of the mRNA template, and cause incorrect amino acids to be incorporated into the growing polypeptide chain
RUO – Research Use Only
General References:
  1. Meyer, R.; Young, L., Armstrong, D. Tobramycin (nebramycin factor 6): in vitro activity against Pseudomonas aeruginosa. Appl Microbiol. 1971, 22, (6), 1147-1151.
  2. Coulter, L.; McLean, R.; Rohde, R.; Aron, G. Effect of bacteriophage infection in combination with tobramycin on the emergence of resistance in Escherichia coli and Pseudomonas aeruginosa biofilms. Viruses. 2014, 3, 6 (10), 3778-3786.
  3. Vicens, Q.; Westhof, E. Crystal structure of a complex between the aminoglycoside tobramycin and an oligonucleotide containing the ribosomal decoding a site. Chemistry & biology. 2002, 9 (6), 747-755.