Aztreonam
Aztreonam
Aztreonam
Aztreonam
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA455410010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAztreonam
TypeAztreonam
CAS78110-38-0
Melting Point227.0°C
Ignition Residue0.1% max.