Carbenicillin disodium salt, 90%
Carbenicillin disodium salt, 90%
Carbenicillin disodium salt, 90%
Carbenicillin disodium salt, 90%
Carbenicillin disodium salt, 90%
Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA455360050
Price (MYR)
1,867.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,867.00
EA
Add to cart
Specifications
Chemical Name or MaterialCarbenicillin disodium salt
TypeCarbenicillin Disodium Salt
CAS4800-94-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
View more
General Description
  • Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
  • In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium
Application
  • Due to its antibacterial activity this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
  • It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
RUO – Research Use Only
General References:
  1. Cole, M. R.; Hobden, J. A.; Warner, I. M. Recycling Antibiotics into GUMBOS: A New Combination Strategy to Combat Multi-Drug-Resistant Bacteria. Molecules 2015, 20 (4), 6466-6487.
  2. Sonopo, M. S.; Venter, K.; Boyle, G.; Winks, S.; Painter, B. M.; Zeevaart, J. R. Carbon-14 radiolabeling and in vivo biodistribution of a potential anti-TB compound. J. Labelled Comp. Radiopharm. 2015, 58 (2), 23-29.