Streptomycin sulfate
Streptomycin sulfate
Streptomycin sulfate
Streptomycin sulfate
Streptomycin sulfate
Streptomycin sulfate, CAS # 3810-74-0, is the sulfate salt form of streptomycin, and an aminoglycoside with antibacterial activity. It also shows a protein synthesis inhibitory activity.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA455341000
Price (MYR)
909.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
909.00
EA
Add to cart
Specifications
Chemical Name or MaterialStreptomycin sulfate
TypeStreptomycin Sulfate
CAS3810-74-0
Health Hazard 1Warning
Health Hazard 2GHS H Statement Suspected of damaging the unborn child. Harmful if swallowed. May cause an allergic skin reaction.
View more
General Description
• Streptomycin sulfate is an aminoglycoside sulfate salt derived from Streptomyces griseus that shows antibacterial activity.
• It inhibits initiation and causes misreading of rRNA in protein synthesis. This compound binds to the S12 protein of the bacterial 30S ribosomal subunit, thereby inhibiting peptide elongation and protein synthesis.
Application
• Streptomycin sulfate can be used to study the mechanisms of streptomycin resistance
• This compound can be used in laboratory applications combined with penicillin to inhibit bacterial contamination in in vitro cell culture
• Streptomycin sulfate shows antibiotic activity against Mycobacterium tuberculosis
• This compound can be used as a protein synthesis inhibitor
• It acts against gram-negative and gram-positive bacteria • Streptomycin sulfate also shows a fungicide activity
• It can be used to remove nucleic acids from a biological extract
RUO – Research Use Only
General References:
  1. Kou, Q. Y.; Li, J.; Zhao, B.; Wu, Z. L. Recovery of streptomycin sulfate from the wastewater using foam fractionation coupled with adsorption separation for reusing sodium dodecyl sulfate. J. Chem. Technol. Biotechnol. 2015, 90 (5), 874-879.
  2. Evans, J. F.; Salvador, V.; George, S.; Trevino-Gutierrez, C.; Nunez, C. Mouse aorta-derived mesenchymal progenitor cells contribute to and enhance the immune response of macrophage cells under inflammatory conditions. Stem Cell Res. Ther. 2015, 6 (56), 1-15.